2',4',6'-Trihydroxypropiophenone ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAT0887-25GEA 1100 CNY
TCIAT0887-25G
2',4',6'-Trihydroxypropiophenone ≥98.0% (通过GC,滴定分析)
2',4',6'-Trihydroxypropiophenone
分子式: C₉H₁₀O₄
MDL编号: MFCD00016456
CAS号: 2295-58-1
EINECS: 218-942-8
https://cn.vwr.com/stibo/web/std.lang.all/49/90/28994990.jpg

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 177.0 to 181.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR