Difluoroacetic anhydride ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Danger
|
TCIAD4164-5GEA
0
CNY
TCIAD4164-5G
Difluoroacetic anhydride ≥98.0% (通过GC)
Difluoroacetic anhydride
分子式:
C₄H₂F₄O₃ 沸点: 127°C |
MDL编号:
MFCD02093315 CAS号: 401-67-2 UN号: 3265 ADR: 8,II |
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...