3,5-Dimethyl-3-hexanol ≥99.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAD1971-5MLEA
0
CNY
TCIAD1971-5ML
3,5-Dimethyl-3-hexanol ≥99.0% (通过GC)
3,5-Dimethyl-3-hexanol
分子式:
C₈H₁₈O 分子量: 130,23 g/mol 沸点: 63°C |
MDL编号:
MFCD00021823 CAS号: 4209-91-0 EINECS: 224-136-7 UN号: 1987 ADR: 3,III |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...