Isobutyltrichlorosilane ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Danger
|
TCIAI0452-25GEA
0
CNY
TCIAI0452-25G
Isobutyltrichlorosilane ≥98.0% (通过GC)
Isobutyltrichlorosilane
分子式:
C₄H₉Cl₃Si 沸点: 140°C 闪点: 37°C |
MDL编号:
MFCD00053185 CAS号: 18169-57-8 EINECS: 242-053-4 UN号: 1747 ADR: 8,II |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...