甲酸香叶酯 >55.0% (通过GC) for perfumery
供应商: TCI EUROPE
|
Warning
|
TCIAG0218-25MLEA
0
CNY
TCIAG0218-25ML
甲酸香叶酯 >55.0% (通过GC) for perfumery
甲酸香叶酯
分子式:
C₁₁H₁₈O₂ 分子量: 182,2594 g/mol 沸点: 92°C 闪点: 96°C |
MDL编号:
MFCD00021047 CAS号: 105-86-2 EINECS: 203-339-4 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 55.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...