Tridecanophenone ≥96.0% (通过GC)
供应商: TCI EUROPE
TCIAT1087-10GEA
0
CNY
TCIAT1087-10G
Tridecanophenone ≥96.0% (通过GC)
Tridecanophenone
分子式:
C₁₉H₃₀O |
MDL编号:
MFCD00059219 CAS号: 6005-99-8 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 40.0 to 43.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...