Undecanophenone ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAU0053-25GEA 445 CNY
TCIAU0053-25G TCIAU0053-500G
Undecanophenone ≥98.0% (通过GC)
Undecanophenone
分子式: C₁₇H₂₆O
分子量: 246,39 g/mol
MDL编号: MFCD00051548
CAS号: 4433-30-1
EINECS: 224-633-9
https://cn.vwr.com/stibo/web/std.lang.all/72/14/28997214.jpg

现在订购

产品技术规格

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR