2,4-双(氯甲基)三甲基苯 ≥98.0% (通过GC)

供应商: TCI EUROPE
Danger

Synonyms: 2,4-双(氯甲基)-1,3,5-三甲苯

TCIAB1994-25GEA 0 CNY
TCIAB1994-25G
2,4-双(氯甲基)三甲基苯 ≥98.0% (通过GC)
2,4-双(氯甲基)三甲基苯
分子式: C₁₁H₁₄Cl₂
分子量: 217,14 g/mol
沸点: 180°C
MDL编号: MFCD00013681
CAS号: 1585-17-7
UN号: 3261
ADR: 8,II
https://cn.vwr.com/stibo/web/std.lang.all/37/53/28933753.jpg

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 108.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR