N-羟基尿烷 ≥85.0% (通过GC,总氮)
供应商: TCI EUROPE
|
Warning
|
Synonyms:
N-羟基氨基甲酸乙酯
TCIAH0618-5GEA
0
CNY
TCIAH0618-5G
TCIAH0618-1G
N-羟基尿烷 ≥85.0% (通过GC,总氮)
N-羟基尿烷
分子式:
C₃H₇NO₃ 沸点: 113°C |
MDL编号:
MFCD00002108 CAS号: 589-41-3 EINECS: 209-648-0 |
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 85.0 % |
Purity (with Total Nitrogen) | min. 85.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...