4'-Aminoacetophenone hydrochloride ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAA0252-25GEA 0 CNY
TCIAA0252-25G
4'-Aminoacetophenone hydrochloride ≥98.0% (通过HPLC,滴定分析)
4'-Aminoacetophenone hydrochloride
分子式: C₈H₁₀ClNO
分子量: 171,6200 g/mol
MDL编号: MFCD00060232
CAS号: 41784-08-1
https://cn.vwr.com/stibo/web/std.lang.all/76/80/28927680.jpg

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR