Funnel holders, SP Bel-Art
供应商: Bel-Art
BELAF182420000EA 476 CNY
BELAF182420000
Funnel holders, SP Bel-Art
支撑架
Non-corrosive polypropylene plastic funnel holder and clamp supports for funnels.
  • For funnels 25 to 152 mm Ø
  • With two bushings for 9,5 or12,7 mm Ø support rods
  • Removable inner ring for smaller funnels