Sponge, NovaCel™
供应商: Micronova
NovaCel™
MICRSP4-533EA 4050 CNY
MICRSP4-533
Sponge, NovaCel™
擦拭巾 海绵
These cleaning sponges are made of PVA (polyvinyl alcohol) and absorb 25 times their weight. Their ultra-clean quality and broad chemical resistance allow them to be used for lapping and polishing in wafer manufacture as well as general wiping applications.
  • Withstand strong disinfectants and steam sterilisation
  • Firm to the touch in their dry state, but once hydrated the material becomes extremely soft and pliant
  • Launderable and autoclavable