D-阿卓糖 99%
供应商: Thermo Scientific
Synonyms:
Alt
ACRO258021000EA
0
CNY
ACRO258021000
D-阿卓糖 99%
D-阿卓糖
分子式:
C₆H₁₂O₆ 溶点: 106…108 °C 存储条件: Ambient |
MDL编号:
MFCD00075620 CAS号: 1990-29-0 EINECS: 217-870-4 |
产品技术规格
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Solubility | (10 % in water) Clear colorless |
HPLC | ≥98.5 % |
Specific optical rotation | +27° to +33° (20°C, 589 nm) (c=2, water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...