2-Hydroxyisophthalaldehyde ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAH1018-5GEA 4525 CNY
TCIAH1018-5G TCIAH1018-1G
2-Hydroxyisophthalaldehyde ≥98.0% (通过GC,滴定分析)
2-Hydroxyisophthalaldehyde
分子式: C₈H₆O₃
存储条件: Refrigerator
MDL编号: MFCD00839420
CAS号: 3328-69-6
https://cn.vwr.com/stibo/web/std.lang.all/10/41/28971041.jpg

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 121.0 to 126.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR