(-)-乙酸冰片酯 ≥70% (通过GC) (contains ca. 20% Isobornyl Acetate)
供应商: TCI EUROPE
Synonyms:
内型-(1S)-1,7,7-三甲基二环[2.2.1]庚烷-2-醇乙酸酯
TCIAB0526-25MLEA
225
CNY
TCIAB0526-25ML
(-)-乙酸冰片酯 ≥70% (通过GC) (contains ca. 20% Isobornyl Acetate)
(-)-乙酸冰片酯
分子式:
C₁₂H₂₀O₂ 分子量: 196,29 闪点: 84°C |
MDL编号:
MFCD00135944 CAS号: 5655-61-8 EINECS: 227-101-4 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 70.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...