Tetrakis(dimethylsilyl)silane ≥95.0% (通过GC)
供应商: TCI EUROPE
|
Danger
|
TCIAT1753-1GEA
0
CNY
TCIAT1753-1G
Tetrakis(dimethylsilyl)silane ≥95.0% (通过GC)
Tetrakis(dimethylsilyl)silane
分子式:
C₈H₂₈Si₅ 存储条件: Refrigerator |
MDL编号:
MFCD01631315 CAS号: 2003-85-2 UN号: 1325 ADR: 4.1,II |
产品技术规格
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...