乙氧甲酰基亚乙基三苯基膦 ≥97.0% (通过HPLC)
供应商: TCI EUROPE
Synonyms:
乙氧甲酰亚乙基三苯基膦
TCIAC1641-5GEA
0
CNY
TCIAC1641-5G
乙氧甲酰基亚乙基三苯基膦 ≥97.0% (通过HPLC)
乙氧甲酰基亚乙基三苯基膦
分子式:
C₂₃H₂₃O₂P 分子量: 362,41 g/mol 存储条件: Freezer |
MDL编号:
MFCD00009160 CAS号: 5717-37-3 EINECS: 227-215-4 |
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Melting point | 159.0 to 163.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...