Tetrabutylphosphonium tetrafluoroborate ≥97.0% (通过滴定分析)

供应商: TCI EUROPE
Danger

TCIAT2006-25GEA 900 CNY
TCIAT2006-25G
Tetrabutylphosphonium tetrafluoroborate ≥97.0% (通过滴定分析)
Tetrabutylphosphonium tetrafluoroborate
分子式: C₁₆H₃₆BF₄P
MDL编号: MFCD00270144
CAS号: 1813-60-1
UN号: 1759
ADR: 8,III
https://cn.vwr.com/stibo/web/std.lang.all/59/47/28995947.jpg

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 97.0 to 100.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR