1-Chloro-2-(trifluoromethoxy)benzene ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAC2091-25GEA 865 CNY
TCIAC2091-25G
1-Chloro-2-(trifluoromethoxy)benzene ≥98.0% (通过GC)
1-Chloro-2-(trifluoromethoxy)benzene
分子式: C₇H₄ClF₃O
沸点: 143°C
MDL编号: MFCD03840206
CAS号: 450-96-4
UN号: 1993
ADR: 3,III
https://cn.vwr.com/stibo/web/std.lang.all/91/47/28939147.jpg

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR