Tetrabutylphosphonium tetraphenylborate ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE

TCIAT2008-25GEA 0 CNY
TCIAT2008-25G
Tetrabutylphosphonium tetraphenylborate ≥98.0% (通过HPLC,滴定分析)
Tetrabutylphosphonium tetraphenylborate
分子式: C₄₀H₅₆BP
分子量: 578,6600 g/mol
MDL编号: MFCD04038112
CAS号: 29089-62-1
https://cn.vwr.com/stibo/web/std.lang.all/59/49/28995949.jpg

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 227.0 to 234.0 deg-C
Solubility in THF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR