Tetraethylphosphonium hexafluorophosphate ≥99.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAT1746-5GEA
0
CNY
TCIAT1746-5G
Tetraethylphosphonium hexafluorophosphate ≥99.0% (通过滴定分析)
Tetraethylphosphonium hexafluorophosphate
分子式:
C₈H₂₀F₆P₂ |
MDL编号:
MFCD01631313 CAS号: 111928-07-5 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...