N-乙酰乙醇胺 ≥95.0% (通过GC)
供应商: TCI EUROPE
|
Danger
|
Synonyms:
N-(2-羟基乙基)乙酰胺
TCIAA0075-500GEA
0
CNY
TCIAA0075-500G
TCIAA0075-25G
N-乙酰乙醇胺 ≥95.0% (通过GC)
N-乙酰乙醇胺
分子式:
C₄H₉NO₂ 分子量: 103,12 g/mol 沸点: 167°C 闪点: 177°C |
MDL编号:
MFCD00002836 CAS号: 142-26-7 EINECS: 205-530-8 |
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...