2-Heptadecylimidazole ≥85.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAH0021-25GEA
0
CNY
TCIAH0021-25G
TCIAH0021-500G
2-Heptadecylimidazole ≥85.0% (通过GC)
2-Heptadecylimidazole
分子式:
C₂₀H₃₈N₂ 分子量: 306,5291 g/mol |
MDL编号:
MFCD00038620 CAS号: 23328-87-2 EINECS: 245-589-7 |
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 85.0 % |
Melting point | 87.0 to 94.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...