3,4-Dimethylcyclohexanol (mixture of isomers) ≥97.0% (通过GC)

供应商: TCI EUROPE

TCIAD0703-25MLEA 0 CNY
TCIAD0703-25ML
3,4-Dimethylcyclohexanol (mixture of isomers) ≥97.0% (通过GC)
3,4-Dimethylcyclohexanol (mixture of isomers)
分子式: C₈H₁₆O
沸点: 189°C
闪点: 78°C
MDL编号: MFCD00001506
CAS号: 5715-23-1
EINECS: 227-211-2
https://cn.vwr.com/stibo/web/std.lang.all/56/41/28975641.jpg

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 % (total of isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR