trans-1,3-Dimethylcyclohexane ≥95.0% (通过GC)
供应商: TCI EUROPE
|
Danger
|
TCIAD1054-10MLEA
0
CNY
TCIAD1054-10ML
trans-1,3-Dimethylcyclohexane ≥95.0% (通过GC)
trans-1,3-Dimethylcyclohexane
分子式:
C₈H₁₆ 沸点: 124°C |
MDL编号:
MFCD00064953 CAS号: 2207-03-6 UN号: 2263 ADR: 3,II |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...