1-(氰甲基)氯化吡啶 ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
|
Danger
|
TCIAC0848-5GEA
0
CNY
TCIAC0848-5G
1-(氰甲基)氯化吡啶 ≥98.0% (通过滴定分析)
1-(氰甲基)氯化吡啶
分子式:
C₇H₇ClN₂ 分子量: 154,60 g/mol |
MDL编号:
MFCD00031933 CAS号: 17281-59-3 EINECS: 241-312-9 UN号: 3439 ADR: 6.1,III |
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...