1-Propylpyridinium chloride ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAP1393-5GEA
0
CNY
TCIAP1393-5G
1-Propylpyridinium chloride ≥98.0% (通过滴定分析)
1-Propylpyridinium chloride
分子式:
C₈H₁₂ClN |
MDL编号:
MFCD06797153 CAS号: 23271-47-8 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...