3-巯基苯甲酸 ≥97.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAM2221-5GEA 1650 CNY
TCIAM2221-5G
3-巯基苯甲酸 ≥97.0% (通过GC,滴定分析)
3-巯基苯甲酸
分子式: C₇H₆O₂S
分子量: 154,1863 g/mol
MDL编号: MFCD00238636
CAS号: 4869-59-4
https://cn.vwr.com/stibo/web/std.lang.all/82/04/28988204.jpg

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %
Melting point 147.0 to 151.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR