2,3,4,6-Tetrafluorobenzyl alcohol ≥98.0% (通过GC)
供应商: TCI EUROPE
TCIAT1942-1GEA
355
CNY
TCIAT1942-1G
TCIAT1942-5G
2,3,4,6-Tetrafluorobenzyl alcohol ≥98.0% (通过GC)
2,3,4,6-Tetrafluorobenzyl alcohol
分子式:
C₇H₄F₄O |
MDL编号:
MFCD03701105 CAS号: 53001-70-0 |
![]() |
产品技术规格
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Freezing point | 16.0 to 20.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...