Cryogenic vial grippers, CryoCeps™
BCISBCS-213MCEA 0 CNY
BCISBCS-213MC BCISBCS-213
Cryogenic vial grippers, CryoCeps™
Gripping tool for moving and sorting frozen cryovials and microtubes. Protects fingers when extracting frozen tubes from cryostorage boxes, CoolRack™ modules or CoolCell® freezing containers.
Unique design grips cryovials by the cap and allows you to lift the tube and move its position. Compatible with internal- or external-threaded cryovials and most microfuge tubes.
Delivery information: The multicolour pack of five includes 1 each of green, pink, purple, orange and blue.