Industrial gloves, Vital 210
供应商: Mapa Professionnel
MAPA34210299EA 0 CNY
MAPA34210299 MAPA34210298 MAPA34210297 MAPA34210296
Industrial gloves, Vital 210
手套
These natural latex gloves with cotton flock lining and textured finish are ideal for minor maintenance tasks and the assembly of small, oil and grease free parts.
  • Freedom of movement
  • Cotton flock lining adds comfort and provides absorption of perspiration
  • Good resistance to many diluted acids
Certifications: EN 388:2016 (1110X), EN ISO 374-1 Type B (KPS), EN ISO 374-5, EN 421; Cat. III