Erlenmeyer flasks, narrow neck, with heavy duty rim, PYREX®
供应商: Corning
PYREX® Corning®
CORN4980-6LEA 0 CNY
CORN4980-6L CORN4980-1XL CORN4980-PACK CORN4980-300 CORN4980-25 CORN4980-125 CORN4980-500 CORN4980-10 CORN4980-50 CORN4980-1L CORN4980-2L CORN4980-4L CORN4980-250
Erlenmeyer flasks, narrow neck, with heavy duty rim, PYREX®
烧瓶 三角烧瓶
PYREX® borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Heavy duty rim flasks have approximate graduations in durable white enamel. Extra large marking space. Heavy, durable top tooling.