Bottles, narrow neck, square,with screw cap
供应商: Corning Life Sciences
Corning®
CORN1396-500EA 0 CNY
CORN1396-500 CORN1396-100 CORN1396-1L CORN1396-250
Bottles, narrow neck, square,with screw cap
玻璃瓶
Made of PYREX® borosilicate glass, with linerless, one-piece, orange PP screw cap with a drip-free pouring ring.
  • Exceptional chemical and thermal resistance
  • Rounded edges allow easy cleaning
  • White graduations and a large marking spot
  • Glass ridge moulded into the bottle indicates full capacity
  • Autoclavable
Square design saves 13 to 20% of shelf space, and slightly convex walls minimise glass-to-glass contact during autoclaving.
Further PP coloured screw caps for ease of sorting and identification, vented membrane screw caps with 0,22 µm PTFE hydrophobic membrane, PBT solid or open top screw caps and ETFE pouring rings are optionally available.