Measuring cylinders, tall form, borosilicate glass, PYREX®
供应商: Corning Life Sciences
PYREX®
CORN3022-25EA 0 CNY
CORN3022-25 CORN3022-50 CORN3022-10 CORN3022-1L CORN3022-2L CORN3022-500 CORN3022-100 CORN3022-250
Measuring cylinders, tall form, borosilicate glass, PYREX®
量筒 Measuring Cylinders
Borosilicate glass.
  • Calibrated to contain (TC In ) at 20 °C (68 °F)
  • Hexagonal base with graduations in white enamel
  • 10 ml cylinder has a funnel top
Use the 100 ml cylinder for ASTM E133, E1272, D86, D216, and D447.
Delivery information: Supplied with bumper guards for cylinders of capacity 25 to 500 ml.