Petri dishes, with cover, PYREX®
供应商: Corning Life Sciences
PYREX®
CORN3160-101EA 0 CNY
CORN3160-150 CORN3160-101 CORN3160-102 CORN3160-152 CORN3160-100 CORN3160-60
Petri dishes, with cover, PYREX®
培养皿 培养皿
PYREX®, borosilicate glass.
These flat, clear dishes will withstand repeated sterilisation (wet or dry). The edges are beaded to provide greater mechanical strength. The bead also provides a means to equally space the side walls of the bottom and cover, thereby reducing the capillary action of the condensed moisture on the sides. They are not affected chemically or thermally by any of the methods commonly employed in laboratories where sterilisation is routinely used. The covered dish is not airtight. The covers are marked in blue enamel and the bottoms in white enamel to make sorting easier. Bottoms also have a triangular enamel reference point for serial dilutions.