Separating funnels, Squibb, PYREX®
供应商: Corning
PYREX® Corning®
CORN6404-500EA 0 CNY
CORN6404-250 CORN6404-60 CORN6404-2L CORN6404-1L CORN6404-500 CORN6404-125
Separating funnels, Squibb, PYREX®
漏斗 分液漏斗
PYREX® Borosilicate glass.
With PTFE stopcock and linear, high-density, closed-bottom polyethylene stoppers which conform to [ST] stopper dimensions.