Oil dilution distilling receiver, with standard taper joints, PYREX®
供应商: Corning
PYREX®
CORN3582EA 0 CNY
CORN3582
Oil dilution distilling receiver, with standard taper joints, PYREX®
蒸馏组件 蒸馏接收器
Distallation receiver to determine the amount of dilution in crankcase oils when gasoline has been used as fuel. Receiver design keeps heat-sensitive materials away from the hot base of the flask. Receiver has a [ST] 24/40 top outer joint and a [ST] 24/40 sidearm inner joint.
Receiver meets ASTM D322 requirements.
Certifications: Receiver meets ASTM D322 requirements.