Screw caps with rubber liner, KIMBLE®
供应商: DWK Life Sciences
KIML73802-15415EA 0 CNY
KIML73802-15415 KIML73802-13415 KIML73800-13415
Screw caps with rubber liner, KIMBLE®
外壳 检测/样品管盖
Phenolic cap material and cemented-in rubber liner are resistant to the effects of repeated autoclaving. Designed from ASTM Specification E982.