Nitrile gloves, ChemSoft
供应商: North by Honeywell
NORHCS046W/LEA 420 CNY
NORHCS046W/L NORHCS046W/XL NORHCS046W/M NORHCS046W/S
Nitrile gloves, ChemSoft
手套
These disposable gloves can be used for fine assemblies of oily parts and for the handling of chemicals.
  • Patterned finish provides a better grip
  • Nitrile offers better mechanical resistance than latex
  • Superior performance when in contact with greasy products
  • Glove is treated to prevent the development of bacteria
  • Non powdered interior ensures the absence of a transfer of cornstarch to foodstuffs, for a lower risk of allergy
Ideal for use in electronics, nuclear industries, packaging and inspection applications, pharma and chemical industries.
Certifications: EN 374, EN 421