Knitted gloves, Light Task Plus, NFD16
供应商: North by Honeywell
NORHNFD16/10XLEA 1375 CNY
NORHNFD16/10XL NORHNFD16/8M NORHNFD16/7S NORHNFD16/9L
Knitted gloves, Light Task Plus, NFD16
These Dyneema®/Lycra® knitted gloves with white polyurethane coating on palm and fingertips are ideal for handling sharp parts in a dry or greasy environment.
  • Tight knit (gauge 13) gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • Ventilated back optimises its breathability
  • Dyneema®/lycra® thread provides outstanding properties against cuts
  • Polyurethane coating at the palm and fingertips provides outstanding abrasion resistance
Please note: These gloves are not suitable for applications involving heat.
Certifications: EN 388:2016 (4X42B); Cat. II