Dry box gloves, Hypalon®
供应商: North by Honeywell
NORH8Y1532A/9QEA 0 CNY
NORH8Y1532A/9Q NORH8Y1532/8H
Dry box gloves, Hypalon®
手套
Hypalon® gloves offer outstanding abrasion and chemical resistance, including resistance to ozone and oxidising chemicals.
  • Naturally soft for greater working comfort
  • White surface for good visibility; will not hide contamination
  • Hand specific or ambidextrous