pH/mV/ISE meter, bench top, H-Series H260G
供应商: Hach
HACHH260GNPEA 16848 CNY
HACHH260GNP
pH/mV/ISE meter, bench top, H-Series H260G
The H-Series laboratory meters are dual-technology pH (non-glass 'ISFET' and traditional glass). They have the ability to accurately measure pH (ISFET or glass), mV, temperature, ORP, and ISE.
  • Large LCD display with LED backlight
  • Up to five point pH calibration
  • Automatic or manual temperature compensation
  • Can be used with either stainless steel, non-glass ISFET probes or traditional glass electrodes with BNC connector
Delivery information: Supplied with USB cable, SmartLogger II software and power supply. Electrodes need to be ordered separately.