pH buffer solutions, BuffAR®, StandARd®
供应商: Avantor®
BuffAR®
MACR0099-04EA 0 CNY
MACR0099-04 MACR0098-04 MACR0097-04
pH buffer solutions, BuffAR®, StandARd®
缓冲(液) 酸碱缓冲液
  • Accurate to within ±0,01 pH at 25 °C
  • Colour coded solutions
Certifications: Traceable to NIST.