Adhesive PCR film and foil seals
供应商: Eppendorf
Masterclear®
EPPE0030132.947EA 4353 CNY
EPPE0030132.947 EPPE0030127.781 EPPE0030127.790
Adhesive PCR film and foil seals
密封件 微孔板封板膜和铝箔
Effective adhesive sealing prevents evaporation loss during PCR.
  • Removal from the plate with no residue
  • Autoclavable (121 °C, 20 min)
  • PCR Film: Sample can be visually monitored through the transparent film
  • PCR Foil: Easily pierced; no sticking of the pipette tip (ideal for automated systems)