Sealing film, self-adhesive for automation
供应商: Brand
BRND701370EA 2350 CNY
BRND701370
Sealing film, self-adhesive for automation
密封件 微孔板封板膜和铝箔
Chemical resistant PE/PP film offers adhesive-free areas over the wells to avoid contamination. Furthermore, it can be pierced easily with a pipette tip (for sample removal). Temperature range: −40 to +90 °C.