Sealing film, self-adhesive for fluorescence measurement
供应商: Brand
BRND701371EA 845 CNY
BRND701371
Sealing film, self-adhesive for fluorescence measurement
密封件 微孔板封板膜和铝箔
Black sealing film made of vinyl is highly suitable for fluorescence measurements because of its light-absorbing characteristics. Temperature range: −40 to +80 °C.