Sealing film, self-adhesive, for cold storage
供应商: Brand
BRND781381EA 1725 CNY
BRND781381
Sealing film, self-adhesive, for cold storage
密封件 微孔板封板膜和铝箔
Aluminium sealing film can be pierced easily with a pipette tip for sample removal. Temperature range: −80 to +120 °C.