4-(2-氨乙基)苯磺酰胺 ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE
Danger

TCIAA1363-25GEA 410 CNY
TCIAA1363-25G
4-(2-氨乙基)苯磺酰胺 ≥98.0% (通过HPLC,滴定分析)
4-(2-氨乙基)苯磺酰胺
分子式: C₈H₁₂N₂O₂S
分子量: 200,26 g/mol
MDL编号: MFCD00010301
CAS号: 35303-76-5
EINECS: 252-501-0
UN号: 3259
ADR: 8,III
https://cn.vwr.com/stibo/web/std.lang.all/92/88/28929288.jpg

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 147.0 to 151.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR