3-Aminopropionitrile ≥98.0% (通过GC,滴定分析) stabilised
供应商: TCI EUROPE
|
Danger
|
Stabilization:
Stabilised with potassium carbonate
TCIAA0408-25MLEA
0
CNY
TCIAA0408-25ML
3-Aminopropionitrile ≥98.0% (通过GC,滴定分析) stabilised
3-Aminopropionitrile
分子式:
C₃H₆N₂ 分子量: 70,10 g/mol 沸点: 185°C 存储条件: Refrigerator |
MDL编号:
MFCD00014820 CAS号: 151-18-8 EINECS: 205-786-0 UN号: 2922 ADR: 8,II |
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...