Methyl-2-methoxy-5-nitrobenzoate ≥98.0% (通过GC)
供应商: TCI EUROPE
TCIAM1437-5GEA
680
CNY
TCIAM1437-5G
Methyl-2-methoxy-5-nitrobenzoate ≥98.0% (通过GC)
Methyl-2-methoxy-5-nitrobenzoate
分子式:
C₉H₉NO₅ 分子量: 211,18 |
MDL编号:
MFCD00673023 CAS号: 34841-11-7 |
![]() |
产品技术规格
Purity (GC) | min. 98.0 % |
Melting point | 98.0 to 103.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...